2-Propanol
2-Propanol
Catalog # EHERC16415200
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)O
- Inchi: InChI=1S/C3H8O/c1-3(2)4/h3-4H,1-2H3
- IUPAC: propan-2-ol
- Packing group: II
Specifications
Specifications
- Description:2-Propanol
- UN No.:1219
- Cat. No.:EHERC16415200
- Formula:C₃H₈O
- Storage temperature:20 °C
- Hazard class description:GHS02, GHS07
- GHS signal:Danger
- Synonym(s):Isopropyl alcohol;Propan-2-ol;2-Propanol
- Molecular weight:60,1
- CAS No.:67-63-0
- Shipping conditions:RT



