Order Entry
China
ContactUsLinkComponent
Microcentrifuge tubes, with or without attached caps
Microcentrifuge tubes, with or without attached caps
  SARS72.690.001
 :  SARSTEDT
Microcentrifuge tubes, with or without attached caps
  SARS72.690.001
 :  SARSTEDT
  • Packed:
    500/bag, 5000/case
  • Type:
    With attached cap
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Natural
  • Pk:
    5.000
PP, translucent.
  • Autoclavable
  • Good chemical resistance
  • Non sterile
For transport, storage and sample processing in the laboratory.