2-丙醇 ≥99.5% (by GC) for spectrophotometry
: TCI
: 仲丙醇, 异丙基醇, 异丙醇, IPA
- :67-63-0
- :(CH₃)₂CHOH
- :MFCD00011674
- :60.1 g/mol
- :14,05208
- :82°C
- :–89 °C
- :0,786 g/cm³ (20 °C)
- :15°C
- :Ambient
- :200-661-7
- :1219
- :3,II
- :01-2119457558-25
: 仲丙醇, 异丙基醇, 异丙醇, IPA
- :67-63-0
- :(CH₃)₂CHOH
- :MFCD00011674
- :60.1 g/mol
- :14,05208
- :82°C
- :–89 °C
- :0,786 g/cm³ (20 °C)
- :15°C
- :Ambient
- :200-661-7
- :1219
- :3,II
- :01-2119457558-25
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
- Absorbance (210nm):max. 0.800
- Absorbance (220nm):max. 0.400
- Absorbance (230nm):max. 0.200
Product Family Options
- 顺序包装类型包装规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



