Order Entry
China
ContactUsLinkComponent
1,2-Dibromoethylene cis- and trans-mixture ≥98.0% (by GC)
Toxic
1,2-Dibromoethylene cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAD0186-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
1,2-Dibromoethylene cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAD0186-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0% (total of isomer)

Specification Test Results

About this item

 :
 :185.85 g/mol
Merck Index:14,00091
Boiling Pt:110°C
Melting Pt:–53 °C
Density:2,268 g/cm³ (0 °C)
EINECS:208-747-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:540-49-8
Formula:C₂H₂Br₂
MDL Number:MFCD00000184