VWR®, Gallium standard solution, 1,000 mg/L Ga in dilute nitric acid AVS TITRINORM standard for AAS
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%