Order Entry
China
ContactUsLinkComponent
1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research
Toxic
Environment
1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAP1826-5G
Supplier:  TCI
CAS Number:  
undefined
Toxic
Environment
1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAP1826-5G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Enzyme inhibition test:
    to pass test

Specification Test Results

About this item

 :
 :198.22 g/mol
Merck Index:14,07214
Melting Pt:102°C
Density:1,259 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:5144-89-8
Formula:C₁₂H₈N₂·1H₂O
MDL Number:MFCD00149973