

N-Methyl-2-pyrrolidone (NMP) for capillary GC analysis
Catalog # SCARME05031000
Supplier: SCHARLAB
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
EINECS:212-828-1
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193