订单输入
China
ContactUsLinkComponent
 
 
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98.0% (通过HPLC,滴定分析)
 :  TCI
 
undefined
 
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98.0% (通过HPLC,滴定分析)
 :  TCI

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    171.0 to 175.0 deg-C
  • Solubility in Acetonitrile:
    very faint turbidity