Order Entry
China
ContactUsLinkComponent
Pyridine ≥99.0% (by GC) sequencing solvent
Flammable
Corrosive
Irritant
Health
Environment
Pyridine ≥99.0% (by GC) sequencing solvent
Catalog # TCIAQ0034-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Environment
Pyridine ≥99.0% (by GC) sequencing solvent
Catalog # TCIAQ0034-100ML
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Peroxide:
    max. 0.001 %

Specification Test Results

About this item

 :
Merck Index:14,07970
Boiling Pt:115°C
Melting Pt:–44 °C
Density:0,983 g/cm³ (20 °C)
Flash Pt:16°C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732