1,1,3,3-Tetramethoxypropane ≥98.0% (by GC) for biochemical research
Catalog # TCIAT2833-5G
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Refractive index n20/D:1.4060 to 1.4090
- TBARS test:to pass test
Specification Test Results
About this item
:
:164.2 g/mol
Boiling Pt:78°C
Density:0,99 g/cm³ (20 °C)
Flash Pt:63°C
Storage Temperature:Refrigerator
EINECS:203-037-2
ADR:3,III
CAS Number:102-52-3
Formula:C₇H₁₆O₄
MDL Number:MFCD00008488