Order Entry
China
ContactUsLinkComponent
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97.0% (by GC, titration analysis)
Irritant
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97.0% (by GC, titration analysis)
Catalog # TCIAD2811-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97.0% (by GC, titration analysis)
Catalog # TCIAD2811-1G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Optical purity (LC):
    min. 98.0 ee%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

About this item

 :
 :444.12 g/mol
 :GHS07
CAS Number:119707-74-3
Formula:C₂₀H₁₂Br₂O₂
MDL Number:MFCD03093629