
9-Aminoacridine hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0619-25G
供应商: TCI
CAS 编号:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
规格
规格测试结果
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Argentmetric Titration):min. 98.0% (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:6.8 to 7.7 %
规格测试结果
关于此项目
:
:248.71 g/mol
默克索引:14,00407
EINECS:205-145-5
ADR:6.1,III
:GHS07
CAS 编号:52417-22-8
化学式:C₁₃H₁₁ClN₂·1H₂O
MDL 编号:MFCD00150071