
p-Tolylhydrazinium chloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAT0319-25G
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Potassium iodate Method):min. 98.0 %
- Solubility in Water:almost transparency
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:158.63 g/mol
Merck Index:14,09542
EINECS:211-295-2
UN:2811
ADR:6.1,III
:GHS06
CAS Number:637-60-5
Formula:C₇H₁₁ClN₂
MDL Number:MFCD00012940