



Hexane (mixture of isomers) ≥99.5% (by GC, sum of isomers + methylcyclopentane) (95% n-Hexane), UltimAR® for liquid chromatography, for organic residue analysis, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:—
- methylcyclopentane)(by GC):≥99.5 %
- ACS - Assay (sum of 5 isomers, total hexanes plus:≥99.5 %
- ACS - Assay (as n-hexane) (by GC, corrected for water):≥95.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0001 %
- ACS - Sulfur Compounds (as S):≤0.005 %
- ACS - Thiophene:Passes Test
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.01 %
- ACS - Water-Soluble Titrable Acid (meq/g):≤0.0003
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 195 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.07
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 - 400 nm:≤0.005
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):≤10 ppb
- FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):≤5 ppb
- FID - Sum of the Peaks (µg/L):≤10
- ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):≤5