Order Entry
China
ContactUsLinkComponent
1,2-Dichlorobutane ≥96.0% (by GC)
Flammable
Irritant
1,2-Dichlorobutane ≥96.0% (by GC)
Catalog # TCIAD0347-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1,2-Dichlorobutane ≥96.0% (by GC)
Catalog # TCIAD0347-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :127.01 g/mol
Boiling Pt:124°C
Density:1,1116 g/cm³ (25 °C)
Flash Pt:27°C
EINECS:210-469-5
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:616-21-7
Formula:ClH₂CCHClCH₂CH₃
MDL Number:MFCD00013676