


Acetonitrile ≥99.5% (by GC) for HPLC
Catalog # TCIAA0793-500ML
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
- Absorbance (210nm):max. 0.200
- Absorbance (220nm):max. 0.050
- Absorbance (230nm):max. 0.020
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:14,00070
Boiling Pt:82°C
Melting Pt:–46°C
Density:0,782 g/cm³ (20 °C)
Flash Pt:10°C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS06
:GHS08
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878