
甲基环己烯酮 ≥98.0% (通过GC)
Catalog # TCIAM1043-25G
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- 包装类型:Glass bottle
- 包装规格:25 g
- 顺序:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:110.16 g/mol
Boiling Pt:197°C
Melting Pt:198…199 °C
Density:0,9791 g/cm³ (10 °C)
Flash Pt:68°C
EINECS:214-769-7
ADR:3,III
:GHS07
CAS Number:1193-18-6
Formula:C₇H₁₀O
MDL Number:MFCD00001581