(±)-1,2-Hexanediol ≥96.0% (by GC)
Supplier: TCI
About this item
Synonyms: Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
- CAS Number:6920-22-5
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00010737
- Molecular Weight:118.18 g/mol
- Boiling Pt:118°C
- Melting Pt:45 °C
- Density:0,9507 g/cm³ (20 °C)
- Flash Pt:122°C
- EINECS:230-029-6
2 Options Available Below
Product Details & Documents
Synonyms: Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
- CAS Number:6920-22-5
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00010737
- Molecular Weight:118.18 g/mol
- Boiling Pt:118°C
- Melting Pt:45 °C
- Density:0,9507 g/cm³ (20 °C)
- Flash Pt:122°C
- EINECS:230-029-6
Certificates
Search by Lot NumberSpecifications for Product Family
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



