Order Entry
China
ContactUsLinkComponent
 
DL-Tryptophan ≥99.0% (by HPLC, titration analysis)
Catalog # TCIAT0540-25G
Supplier:  TCI
CAS Number:  
undefined
DL-Tryptophan ≥99.0% (by HPLC, titration analysis)
Catalog # TCIAT0540-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in HCl:
    almost transparency

Specification Test Results

About this item

 :
 :204.23 g/mol
Melting Pt:289…290 °C
Density:1,349 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-194-9
CAS Number:54-12-6
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064339