Burettes, straight stopcock, Schellbach, with USP individual or DAkkS calibration certificate, BLAUBRAND®
Supplier: BRAND
Certificates
About this item
Borosilicate glass 3.3, class AS.
- Calibrated to deliver 'TD, Ex', conformity certified
- Schellbach stripe, ring marks at major graduations
- Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm