Order Entry
China
Orders LinkContactUsLinkComponent
VWR®, Floor Mat Frames for PureStep Mats
VWR®, Floor Mat Frames for PureStep Mats
Supplier:   VWR Collection
VWR®, Floor Mat Frames for PureStep Mats
Supplier:  VWR Collection
Certificates
VWR®, Floor Mat Frames for PureStep Mats
VWR®, Floor Mat Frames for PureStep Mats
8 Options Available Below
Product Details & Documents
These frames are designed to be used with PureStep mats to create a system, that is ideal for any critical area, and does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, polystyrene base material, combined with a non skid backing, that holds the mats in place on almost any surface.
  • Protects floors from adhesive residue
  • Reduces the risk of slips and falls
  • Non slip parts made from polymer coated, polyester textile
  • Naturally antimicrobial, mould and mildew resistant
Ordering information: Adhesive mats are sold separately.
Product Family Options
Product Information
  • Colour
    For
    L×W
    Thickness
    Availability
    Price
  • Mat frame
    White
    457×914 mm
    508×965 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥2,465.00
    ¥2,465.00
    1 * 1 件
       ¥2,465.00
  • Mat frame
    White
    457×1143 mm
    508×1194 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥3,523.00
    ¥3,523.00
    1 * 1 件
       ¥3,523.00
  • Mat frame
    White
    609×914 mm
    660×965 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥3,807.79
    ¥3,807.79
    1 * 1 件
       ¥3,807.79
  • VWRI115-2471
    Mat frame for 609.6×762 mm mats, white
    Mat frame for 609.6×762 mm mats, white
    635×787,4 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥1,269.00
    ¥1,269.00
    1 * 1 件
       ¥1,269.00
  • Mat frame
    White
    660×1143 mm
    711×1193 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥3,913.00
    ¥3,913.00
    1 * 1 件
       ¥3,913.00
  • Mat frame
    White
    914×914 mm
    965×965 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥4,703.00
    ¥4,703.00
    1 * 1 件
       ¥4,703.00
  • Mat frame
    White
    914×1140 mm
    965×1194 mm
    3,2
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥5,061.00
    ¥5,061.00
    1 * 1 件
       ¥5,061.00
  • Mat frame
    White
    914×1524 mm
    965×1574 mm
    3,2 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥8,660.00
    ¥8,660.00
    1 * 1 件
       ¥8,660.00