5-Fluoro-2'-deoxyuridine (floxuridine) 99+%
: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
:50-91-9
:
:C₉H₁₁FN₂O₅
:MFCD00006530
:246.19 g/mol
:148…153 °C
:Refrigerator
:200-072-5
:2811
:6.1,III
:GHS06
:GHS08
- Environmentally Preferable:
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Melting point:145 to 153 °C
- Loss on drying:≤0.5 % (60 °C, 4 hrs) (vacuum)
- Sulfated ash:≤0.1 %
- HPLC:≥99.0 %
- Specific optical rotation:+26° to +39° (20 °C, 589 nm) (c=1, Water)
- Heavy metals (as Pb):≤20 ppm