Antibacterial wipes for general applications, NanoTech micro
NanoTech micro cloths not only wipes away bacteria, they kill them by direct contact due to the nano particles of silver embedded in the microfibre structure.
- Microfibres and and nano silver particles, reusable
- Efficiently prevents bacterial growth: Removes 99,9% bacteria or germs during wiping (certified by BMA)
- Keeps antibacterial properties even after several hundred wash cycles
- Quick and easy handling due to low friction
- Colour coding system allows cleaning process to meet hygiene standards
The cloth can be used everywhere where hygiene is a priority – e.g. hospitals, care homes, kindergarten, restaurants and fast food environments.