Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Column Format:Analytical
- Ø int.:0,075 mm
- Particle Size:2,6 µm
- Pore size:150 Å
- Length:150 mm
- Phase:C4
- Pk:1
Specifications
About this item
Based on Core Enhanced Technology, these columns provide fast resolution separation of peptides and proteins and generate only 50% of the back pressure associated with columns packed with sub 2 µm materials.
- Fast biomolecule separation
- Rugged, easy to use analytical- and nano-scale columns