
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid) 99% for biochemistry
ACRO172601000
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- UV:(0.1 M in water) 1 cm cell
- pH:5 to 6 (10 % w/v at 20°C)
- Titration with NaOH:≥98.5 %
:
:237…239 °C
:252.33 g/mol
:240-198-8
:GHS07
:Ambient
:16052-06-5
:C₉H₂₀N₂O₄S
:MFCD00006160