TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 0.5 mol/l in aqueous solution, Supelco®
Catalog # SUPL646547-10X1ML
Supplier: SUPELCO
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:286.65 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:51805-45-9
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469