

Methotrexate hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:133073-73-1
:
Formula:C₂₀H₂₂N₈O₅·xH₂O
MDL Number:MFCD00150847
:454.44 g/mol (anhy)
Melting Pt:195 °C (dec.)
Storage Temperature:Refrigerator
EINECS:200-413-8
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Water:≤13 % (K.F.)
- HPLC:≥98.5 %
- Specific optical rotation:+19° to +24° (20°C, 589 nm) (c=1, 14 g/l Na2CO3) (on anhydrous substance)