Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate, Ultra Pure Grade for biochemistry
Catalog # VWRC0634-1KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- A250/A260:0.79 – 0.9
- A280/A260:0.18 – 0.22
- Em (258 nm, phosphate buffer, pH 7.0):>11000
- Heavy metals:<0.002%
Specification Test Results
About this item
:
Melting Pt:>300 °C
:391.2 g/mol (anhy)
Storage Temperature:Refrigerator
EINECS:224-961-2
CAS Number:4578-31-8
Formula:C₁₀H₁₂N₅Na₂O₇P·H₂O
MDL Number:MFCD00065023