
Naphthol AS-BI phosphate 93%
Catalog # ACRO415310010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥91 %
- GC:≥88 %
Specification Test Results
About this item
:
:452.1926 g/mol
EINECS:217-645-0
:GHS07
Storage Temperature:Refrigerator
CAS Number:1919-91-1
Formula:C₁₈H₁₅BrNO₆P
MDL Number:MFCD00004058