
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%
:51805-45-9
:
:C₉H₁₆ClO₆P
:MFCD00145469
:286.65 g/mol
:Refrigerator
:3261
:8,II
:GHS05
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Solubility:(50mg/ml water) Clear colorless
- Titration with NaOH:≥97.5 %