
2-Chloro-N,N-diethylacetamide 98%
Catalog # ACRO351850050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4720 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
:149.62 g/mol
Boiling Pt:121…122 °C (12 mmHg)
Density:1.089 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:219-019-2
UN:2810
ADR:6.1,II
:GHS07
CAS Number:2315-36-8
Formula:C₆H₁₂ClNO
MDL Number:MFCD00000928