Order Entry
China
Orders LinkContactUsLinkComponent
Adapters, extension
Adapters, extension
Supplier:   COWIE
Adapters, extension
Supplier:  COWIE
Certificates
Adapters, extension
Adapters, extension
7 Options Available Below
Product Details & Documents
NPT extension adapters are manufactured in glass filled PTFE for improved mechanical strength.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
Male/female NPT extension adapters optimise the use of NPT entry ports by the provision of additional port sizes and raising the height of the entry port from the lid. This gives even more flexibility of assembly and enables a greater range of fittings to be used - this is especially the case for 60mm lids with multiple ports where the use of an extension adapter allows the use of a central stirrer guide.
Product Family Options
Product Information
  • Availability
    Price
  • COWIPC713.050050GF
    ¹/₂" NPT Male / ¹/₂" NPT Female, for shafts max. 14 mm Ø
    ¹/₂" NPT Male / ¹/₂" NPT Female, for shafts max. 14 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.050075GF
    ¹/₂" NPT Male / ³/₄" NPT Female, for shafts max. 14 mm Ø
    ¹/₂" NPT Male / ³/₄" NPT Female, for shafts max. 14 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.025050GF
    ¹/₄" NPT Male / ¹/₂" NPT Female
    ¹/₄" NPT Male / ¹/₂" NPT Female
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.075075GF
    3/4" NPT Male / 3/4" NPT Female, for shafts max.18 mm Ø
    3/4" NPT Male / 3/4" NPT Female, for shafts max.18 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.075100GF
    3/4" NPT Male / 1" NPT Female, for shafts max. 18 mm Ø
    3/4" NPT Male / 1" NPT Female, for shafts max. 18 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.100100GF
    1" NPT Male / 1" NPT Female, for shafts 24 mm Ø
    1" NPT Male / 1" NPT Female, for shafts 24 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • COWIPC713.100150GF
    1" NPT Male / 1½" NPT Female, for shafts max. 24 mm Ø
    1" NPT Male / 1½" NPT Female, for shafts max. 24 mm Ø
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件