关于此项目
PCR nucleotide mix dNTP set (2 mM, 10 mM, or 25 mM each A,C,G,T) for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready to use solutions in multiple formats
- Functionally tested for long PCR and sequencing