Order Entry
China
ContactUsLinkComponent
Flow indicator, SP Bel-Art
Flow indicator, SP Bel-Art
  BELAH199350000
Flow indicator, SP Bel-Art
  BELAH199350000
  • Maximum Flow Rate:
    100 - 1000  ml/min
  • Description:
    Flow indicator
  • Material:
    PC
  • Pk:
    1
  • Tube Ø:
    6,4 - 9,5 mm
Flow indicator with polypropylene non returning valve.