Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
Buffer, TE solution (TRIS-EDTA buffer)
Print
Buffer, TE solution (TRIS-EDTA buffer)
Certificates
Supplier:
PROMEGA
Buffer, TE solution (TRIS-EDTA buffer)
Supplier:
PROMEGA
Certificates
Return to Top
Buffer, TE solution (TRIS-EDTA buffer)
Product Family Options
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Certificates
Search by Lot Number
Product Family Options
Product Information
Pack type
Pk
Availability
Price
PROMV6232
TE buffer solution (TRIS-EDTA buffer)
TE buffer solution (TRIS-EDTA buffer)
Glass bottle
500 mL
Log in
for availability
Log in
for availability
1 * 500 毫升
-
+
Inquire For Price
1 * 500 毫升
-
+
Inquire For Price
PROMV6231
TE buffer solution (TRIS-EDTA buffer)
Certificates
TE buffer solution (TRIS-EDTA buffer)
Glass bottle
100 mL
Log in
for availability
Log in
for availability
1 * 100 毫升
-
+
Inquire For Price
1 * 100 毫升
-
+
Inquire For Price