¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
Buffers
Cloning Buffers
Buffer, TE solution (TRIS-EDTA buffer)
Buffer, TE solution (TRIS-EDTA buffer)
TE buffer solution (TRIS-EDTA buffer), Glass bottle, 100 mL
INVT12090015
Marketsource
:
INVITROGEN
Buffer, TE solution (TRIS-EDTA buffer)
INVT12090015
:
INVITROGEN
Marketsource
:
Buffer, TE solution (TRIS-EDTA buffer)
TE buffer solution (TRIS-EDTA buffer), Glass bottle, 100 mL
:
1 * 100 毫升
-
+
Pack type:
Glass bottle
Pk:
100 mL
Description:
TE buffer solution (TRIS-EDTA buffer)