Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 mL
目录编号 PROMV4231
供应商: PROMEGA
Jump to:
Product Details & Documents
This EDTA solution is a chelator of divalent cations and is suitable for biochemistry and molecular biology applications
- Available in 0,5 M or 10 mM concentrations
- 10 mM concentration also provided as part of Maxwell® HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)
It is supplied as a solution in deionized water.
规格
规格
- Pack type:Glass bottle
- Pk:100 mL
- Description:EDTA buffer solution pH 8,0 (0,5 mol/l)