Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
Catalog # SIALP3813-10PAK
Supplier: SIGMAALDRICH
CAS Number:
Specifications
- Pk:10
- Description:PBS buffer solution (phosphate buffered saline) pH 7,4
- M-Clarity quality level:MQ200