Order Entry
China
ContactUsLinkComponent
Buffer, PBS solution 10X concentrate (phosphate buffered saline), Sigma-Aldrich®
Buffer, PBS solution 10X concentrate (phosphate buffered saline), Sigma-Aldrich®
Catalog # SIALP5493-4L
Supplier:  SIGMAALDRICH
CAS Number:  
Buffer, PBS solution 10X concentrate (phosphate buffered saline), Sigma-Aldrich®
Catalog # SIALP5493-4L
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pk:
    4 L
  • Description:
    PBS buffer solution 10X concentrate (phosphate buffered saline)
  • M-Clarity quality level:
    MQ200

Specifications

About this item