Order Entry
China
ContactUsLinkComponent
 
Irritant
1,3-Dichloroisoquinoline 97%
Catalog # ACRO382410010
CAS Number:  
undefined
Irritant
1,3-Dichloroisoquinoline 97%
Catalog # ACRO382410010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    120 to 125 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :198.05 g/mol
Melting Pt:120…122 °C
ADR:6.1,III
 :GHS07
Storage Temperature:Ambient
CAS Number:7742-73-6
Formula:C₉H₅Cl₂N
MDL Number:MFCD00034750