Ethylmagnesium bromide 40% 3 M in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:925-90-6
:
Formula:C₂H₅BrMg
MDL Number:MFCD00000043
:133.27 g/mol
Density:1.03 g/cm³
Flash Pt:–40 °C
EINECS:213-127-3
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark brown Solution
- Titration Grignard:2.80 to 3.20 M