Order Entry
China
Orders LinkContactUsLinkComponent
Storage tubes, 2D barcoded, Matrix™

Storage tubes, 2D barcoded, Matrix™

   Sustainable Options Available
Storage tubes, 2D barcoded, Matrix™
   Sustainable Options Available
Certificates
Storage tubes, 2D barcoded, Matrix™

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Permanently attached, laser-etched 2D barcodes on the bottom of each tube allow you to scan and decode tubes at once without removing them from the rack and allow data to be associated with each tube.

  • The 8×12 format enables the filling of 8, 12 or 96 tubes at once
  • Tubes come in latch racks for added security and protection of samples
  • Built-in error correction ensures that barcodes will never be misread
  • 100% quality control checks that every code is unique and readable
  • Solid, one-piece construction enables storage down to cryogenic temperatures

Septum tubes made from Virgin Class VI medical grade PP, autoclavable. The permanently installed DuraSeal® septa enable long-term storage needs at temperatures down to –150 °C.

Glass tubes are ideal for applications requiring enhanced chemical resistance. For samples in DMSO, chlorinated solvents and other harsh chemicals and applications where newly synthesised compounds need to be dried-down or lyophilised and subsequently resolubilised by hand or via automation.

Screwtop tubes and caps made from Virgin Class VI medical grade PP. O-rings made from silicone. These tubes are ideal for sample storage in virtually any laboratory condition, including the vapour phase of liquid nitrogen. They are available with or without caps and can withstand temperatures from –180 to +121 °C.

All tubes and trays are supplied free from DNA, RNAse, DNAse, endotoxins and cytotoxins. Tubes should be tested under individual laboratory conditions.

Specifications for Product Family

Specifications

  • Catalog No:
  • MATR3790
  • MATR3791
  • MATR3744
  • MATR3744-BR
  • Environmentally Preferable:
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
  • NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement

Specifications

Product Family Options

Septum tubes

Product Information

  • Capacity
    Base
    Cap included
    Cap colour
    Sterile
    Packed
    Availability
    Price
  • MATR3790
    2D barcoded tubes
    2D barcoded tubes
    1,40 ml
    V bottom
    No
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 件
    1 * 1.000 件

    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    MATR3790
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • MATR3791
    2D barcoded tubes
    2D barcoded tubes
    1,40 ml
    V bottom
    No
    -
    10 latch racks of 96 tubes
    Log in for availability
    Log in for availability
    1 * 960 件
    1 * 960 件

    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    MATR3791
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure

Screwtop tubes with caps in non barcoded racks

Product Information

  • Capacity
    Base
    Cap included
    Cap colour
    Sterile
    Packed
    Availability
    Price
  • MATR3744
    2D barcoded screwtop tubes with caps, 5 racks of 96 tubes per case, sterile
    2D barcoded screwtop tubes with caps, 5 racks of 96 tubes per case, sterile
    0,50 ml
    V bottom
    Yes
    Colourless
    Log in for availability
    Log in for availability
    1 * 480 件
    1 * 480 件

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MATR3744
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact

Screwtop tubes with caps in barcoded racks

Product Information

  • Capacity
    Base
    Cap included
    Cap colour
    Sterile
    Packed
    Availability
    Price
  • MATR3744-BR
    2D barcoded screwtop tubes with caps, 5 latch racks of 96 tubes per case, sterile
    2D barcoded screwtop tubes with caps, 5 latch racks of 96 tubes per case, sterile
    0,50 ml
    V bottom
    Yes
    Colourless
    Log in for availability
    Log in for availability
    1 * 480 件
    1 * 480 件

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MATR3744-BR
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact

Racks and lids

Product Information

  • Catalog #
    Colour
    Description
    For
    Availability
    Price
  • White
    Latch rack, empty, plastic
    1,4 ml tubes
    Log in for availability
    Log in for availability
    1 * 10 件
    1 * 10 件 - ¥0.00
    ¥0.00
    1 * 10 件
       ¥0.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.