


Chlorobenzene ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACR4426-08
Supplier: MACRON AVANTOR BRAND
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- ACS - Assay (C₆H₅Cl) (by GC, corrected for water):≥99.5 %
- ACS - Color (APHA):≤30
- ACS - Residue after Evaporation:≤2 ppm
- ACS - Titrable Acid (meq/g):≤0.004 %
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.03 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 325 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:≤0.005
Specification Test Results
About this item
:
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–45 °C
Density:1.11 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
:Management of Change (MOC) category = N
REACH:01-2119432722-45
:GHS02
:GHS07
:GHS09
CAS Number:108-90-7
Formula:C₆H₅Cl
MDL Number:MFCD00000530