Order Entry
China
ContactUsLinkComponent
Dichloromethane ≥99.9% stabilised, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Health
Irritant
Dichloromethane ≥99.9% stabilised, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH485-80
CAS Number:  
undefined
Health
Irritant
Dichloromethane ≥99.9% stabilised, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH485-80
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • ACS - Assay (CH₂Cl₂ including stabilizer):
    ≥99.9 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Free Halogens:
    Passes Test
  • ACS - Residue after Evaporation:
    ≤0.0001 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Water (H₂O)(by coulometry):
    ≤0.01 %
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 233 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm:
    ≤1.0
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.005
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 - 400 nm:
    ≤0.003
  • Liquid Chromatography Suitability Absorbance:
    Passes Test
  • ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):
    ≤5
  • Sum of the Peaks (ng/L):
    ≤10
  • FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):
    ≤5
  • Sum of the Peaks (µg/L):
    ≤10
  • Preservative - Methanol (CH₃OH):
    ≥400 ppm
  • Preservative - Amylenes:
    ≥8 ppm

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881