Dimethyl sulphoxide ≥99.9% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
供应商: MACRON AVANTOR BRAND
证书
关于此项目
CAS 编号:67-68-5
:
化学式:(CH₃)₂SO
MDL 编号:MFCD00002089
:78.14 g/mol
默克索引:13,03285
沸点:189 °C (1013 hPa)
熔点:18.5 °C
密度:1.101 g/cm³ (20 °C)
闪点:87 °C
储存温度:Ambient
EINECS:200-664-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
规格
规格测试结果
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay ((CH₃)₂SO) (by GC, corrected for water):≥99.9 %
- Appearance (clear, colorless liquid):Passes Test
- ACS - Residue after Evaporation:≤0.01 %
- ACS - Titrable Acid (meq/g):≤0.001
- ACS - Water (H₂O)(by coulometry):≤0.1 %