Choline chloride ≥98.0%, high purity
Catalog # VWRC0448-1KG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Bottle, amber
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- 1,4-Dioxane:Pass
- Arsenic:0.0002%
- Heavy metals:0.001%
- Moisture:0.5%
- pH (25%, Water) @25 °C:4.5 − 7.5
- Purity (Anhydrous):98.0%
- Residue on ignition:0.05%
Specification Test Results
About this item
:
:139.62 g/mol
Merck Index:13,02226
Melting Pt:244−247 °C
Melting Pt:244…247 °C
Density:∼1.205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-655-4
CAS Number:67-48-1
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721