Order Entry
China
ContactUsLinkComponent
 
Maxi submarine electrophoresis unit, HE 99X
Catalog # GEHEHE99X-15-1.5
Supplier:  GE HEALTHCARE
undefined
Maxi submarine electrophoresis unit, HE 99X
Catalog # GEHEHE99X-15-1.5
Supplier:  GE HEALTHCARE

Specifications

  • Model:
    HE 99X
  • Description:
    Max submarine electrophoresis unit, complete (incl. basic unit, 15×20 cm gel casting kit, 1×1,5 mm thick 15-well comb, and comb back)
  • Buffer volume:
    1200 ml
  • Gel size W×L:
    150×200 mm
  • Max. power:
    20 W
  • Sample capacity:
    60 samples
  • Unit W×L×H:
    182×360×140 mm
  • Pk:
    1

Specifications

About this item

HE 99X Maxi submarine unit offers a variety of gel lengths and comb sizes so that the gel can be run exactly based on the need.

  • Versatile submarine unit is ideal for any molecular biology lab
  • Run as many as 60 samples at once

Cast gels 15 cm wide × 10, 15, or 20 cm long, with one or two combs. Comb choices range from a preparative comb, for a 1 ml fragment purification sample, to a 30-well comb for high-throughput PCR screening.

Every design detail contributes to reliability and durability with seamless molded high impact plastic buffer tray; banana plugs safe from corrosion; a guard to protect the platinum electrodes; electrode carrier for simple electrode replacement; a rugged molded lid with safety guards to protect against electrical hazard; a large diameter bubble level and leveling feet; strain-relieved high voltage power cords; and red and black details on running trays and buffer chamber for orientation.