Media supplements for dehydrated culture media
Catalog # MERC1.09875.0010
Supplier: MERCK MILLIPORE
CAS Number:
Specifications
- Pk:10 Vial
- Description:Bacillus cereus selective supplement
Specifications
About this item
The isolation of microorganisms often requires the use of selective media. The use of freeze-dried supplements ensures an optimal and standardised medium preparation and thus optimised performance of the selective medium.
- No aerosols released upon opening, therefore minimising inhalation of toxic agents
- User friendly screw caps which guarantee easy and safe opening
- Standard volumes make preparation easier