Order Entry
China
ContactUsLinkComponent
Corning® Cell-Tak™ Cell and tissue adhesive
Corning® Cell-Tak™ Cell and tissue adhesive
Catalog # BDAA354240
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® Cell-Tak™ Cell and tissue adhesive
Catalog # BDAA354240
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Mytilus edulis
  • Description:
    Corning® Cell-Tak™ cell and tissue adhesive
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Molecular weight:
    110 - 140 kD
  • Formulation:
    As a liquid in 5% acetic acid
  • Quality control:
    Dopa: protein ratio >0,05
    Tested for ability to promote cell attachment and spreading of BHK-21 and U937 cells
    Tested and found negative for bacteria, fungi, and mycoplasma
  • Storage and stability:
    Stable for at least three months at 2 to 8 °C. Do not freeze.

Specifications

About this item

Corning® Cell-Tak™ Cell and Tissue Adhesive is a formulation of polyphenolic proteins extracted from Mytilus edulis (marine mussel). These proteins are the key components of the glue secreted by the mussel to anchor itself to solid substrates in the marine environment. Corning® Cell-Tak™ is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, PTFE, and biological materials. It can simplify the manipulation of biological samples for a number of in vitro techniques, including in situ hybridisation, immunoassays, microinjection, immunohistochemistry, and establishing primary cells in culture. Corning® Cell-Tak™ is biocompatible and demonstrates no species specificity.