Bomb sampler, Wheaton®
Bomb sampler, Stainless steel, 500 ml
Catalog # WHEA885200
Supplier: DWK LIFE SCIENCES
Jump to:
Product Details & Documents
An efficient and unique composite sampling device made of stainless steel.
- Cylindrical chamber
- Weighted plunger that seals the chamber
- Cable attachments for suspending the apparatus and activating the sampling device
Suitable for collecting liquid from top, bottom or any intermediate level of storage tanks, tank cars, lagoons, ponds and wells.
Certifications: EPA composite sampling methods 'Test Methods for Evaluating Solid Waste 1.1.4.1'.
Specifications
- Capacity:500 ml
- Description:Bomb sampler
- Material:Stainless steel